Would you like to log in again?
VWR® Ratchet Headgear with Face Shield
Supplier: VWR
VWR®
I-1BG-01 I-1
76336-852CS 579.58 USD InStock
76336-852 76336-850
VWR® Ratchet Headgear with Face Shield
Face Shields
The VWR® Ratchet Headgear with Face Shield provides full face protection from debris and splash hazards. Ideal for use in many environments ranging from the lab to industrial applications.
  • Lightweight design
  • Ratchet headgear features padded headband for better fit and comfort
  • Face shield features universal slot design compatible with many different headgear styles
  • Fits comfortably with respirator or eyewear
  • Impact-resistant, scratch-resistant polycarbonate shield
Face shields should be worn with ANSI approved eyewear.
Certifications: ANSI Z87.1 certified
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty