Would you like to log in again?
SULFINERT REGULATOR DUAL STAGE CGA 320
Supplier: Avantor
Sulfinert Regulator Dual Stage Cga 320^ Regulator, dual stage, cem continuous emission monitoring, Material: Sulfinert-Treated, Fitting : CGA 320 CO2, Outlet Gauge: 30 in, 0 to 200 psig (0-1,379 kPa), Outlet Pressure: 0 to 150 psig (0-1,034 kPa), Environmental stack and gas emission standards
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SULFINERT REGULATOR DUAL STAGE CGA 320 76336-946   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty