Would you like to log in again?
Sani-Cloth® Bleach Germicidal Disposable Wipes, PDI®
Supplier: PDI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sani-Cloth®
PDIP25784 PDIP7007P PDIH58195 PDIP54072 PDIP700RF PDIU26595
76337-092CS 251.95 USD InStock
76337-092 76337-108 76337-062 76337-024 76337-020 76337-022
Sani-Cloth® Bleach Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Ideal for disinfecting high risk areas endemic with multi-drug resistant organisms, Clostridium, difficile spores, and Norovirus.
  • Compatible with a broad range of surfaces and equipment in healthcare
  • Up to 2× greater compatibility vs competitors with materials used in healthcare
  • 20% extended equipment life compared to competitors
  • No visible degradation on stainless steel, ABS plastic, brass alloy, and more
Effective against 50 micro organisms in four minutes, including the following MDRO's, blood borne pathogens, and viruses: C.difficile spores, Acinetobacter baumannii, Klebsiella pneumoniae, Candida albicans, ESB-resistant E. coli, ESB-resistant Klebsiella pneumoniae, Norovirus, MRSA, VRE, VRSA, HIV, HBV, and HCV.
Wipes may be used on hard, non porous surfaces in healthcare facilities such as bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to see a full listing of use sites.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty