Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

ProPette™ LE Adjustable Pipettors, MTC Bio

Supplier: MTC Bio
Propette™ LE
P5200-5M P5200-100U P5200-1M P5200-20U P5200-10M P5200-50U P5200-2U P5200-200U P5200-300U P5200-10U
76337-016EA 386.27 USD InStock
76337-016 76337-126 76337-116 76337-018 76337-128 76337-118 76337-122 76337-124 76337-130 76337-120
ProPette™ LE Adjustable Pipettors, MTC Bio
Pipettes Repetitive Pipettors
The ProPette LE™ is the next generation of high precision liquid handling instruments from MTC Bio. This addition to the Propette family features improved actuation forces and a nearly 20% reduction in overall weight.

  • Adjustable with Roto-Lock™
  • Light weight with reduced forces
  • Full volume range (0.1 µl to 10 ml)
  • Superior accuracy and reproducibility
  • Outstanding user comfort

Every aspect of accuracy, reliability and user comfort has been carefully reviewed in designing these high precision pipettes.  The result is an advanced pipette that exhibits superior accuracy and ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.

Certifications: CE marked.

Delivery information: Includes calibration key.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty