Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FastCounting™ Cell Counting Kits, MP Biomedicals

Supplier: MP Biomedicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

092690132 092690131
76337-148EA 226.21 USD InStock
76337-148 76337-146
FastCounting™ Cell Counting Kits, MP Biomedicals
Assays Cellular Assays
The FastCounting™ cell counting kits provide a sensitive and convenient method to determine cell viability in cell proliferation and cytotoxicity assays.

  • Benefits of using FastCounting cell kit
  • Highest sensitivity dye for cell viability
  • Lowest cytotoxicity among all tetrazolium reagents
  • Simple procedures without thawing reagents
  • Long shelf life and easy storage
  • Store at –20 °C long-term

The kit uses a highly sensitive tetrazolium dye with low cytotoxicity to quantify the number of living cells. Sufficient for 100 tests. The FastCounting cell counting kit is stable for over one year at 2 to 8 °C when protected from light.

Caution: Avoid repeated thawing and freezing, which can cause an increase in background.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty