Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FastMitophagy™ Detection Kit, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

092690201
76337-150EA 694.85 USD InStock
76337-150
FastMitophagy™ Detection Kit, MP Biomedicals
Assays Cellular Assays
The FastMitophagy™ detection kit is specifically designed for mitophagy detection in mammalian cells.

  • Store at 0 to 5 °C with protection from light

The kit is composed of a mitophagy dye, a reagent for detection of mitophagy, and lysosome dye. Reagents are sufficient for 5×96 well plates or 25 assays when using a 35 mm dish. Mitophagy dye initially accumulates in intact mitochondria and becomes immobilized via chemical bonding, resulting in a very weak fluorescence due to the influence of the surrounding environment.

When mitophagy is induced, the damaged mitochondria fuses to lysosome and the Mitophagy dye emits a high fluorescence. Lyso dye is used to confirm the fusion of Mitophagy dye - labeled mitochondria and lysosome.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty