Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody [clone: HB01]

Supplier: MP Biomedicals
08634801
76337-158EA 435 USD InStock
76337-158
Anti-IgG Mouse Monoclonal Antibody [clone: HB01]
Antibodies
Anti-Human Hemoglobin Monoclonal Antibody from Mouse (10 mM Phosphate Buffer, pH 7.4, containing 0.9% NaCl and 0.02% NaN3). Hemoglobin’s chief physiological function is to transport oxygen from the lungs or gills to the tissues where it is released it for cellular respiration.

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone: HB01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty