Would you like to log in again?
Bovine Brain Extract, MP Biomedicals
Supplier: MP Biomedicals
092850550
76337-166EA 67.23 USD InStock
76337-166
Bovine Brain Extract, MP Biomedicals
Cell Culture Supplements and Additives
Bovine Brain Extract (BBE) is made from fresh, unfrozen pituitary glands from young, healthy New Zealand cattle less than 30 months old.
It is formulated to be used from 0.1 to 0.2% in the final serum-free medium.
Caution: Do not thaw and refreeze.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty