Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Puro™ III T and Geno™ II T All-In-One Lab Water Systems with Integral 35 L Tank, Avidity Science™

Supplier: Avidity Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

GENOIIT20UV PUROIIIT10 PUROIIIT20 GENOIIT20 GENOIIT10UV PUROIIIT20UV GENOIIT10 PUROIIIT10UV
76337-260EA 5849.53 USD InStock
76337-260 76337-258 76337-254 76337-266 76337-256 76337-268 76337-262 76337-264
Puro™ III T and Geno™ II T All-In-One Lab Water Systems with Integral 35 L Tank, Avidity Science™
Water Purification Systems Deionizers
Puro™ III T and Geno™ II T are small in size; big on benefits. Providing RO or DI lab water to startup facilities.

  • Innovative design with 35 L integrated tank
  • Low energy consumption (<60 W)
  • Unique disposable RO maintains peak efficiency
  • Vibration-free system
  • Mercury-free UV option
  • Unique disposable RO maintains peak efficiency
  • Consumables changed in seconds
  • Ideal replacement of stills
  • Easy-to-install, low maintenance
  • Minimal water wastage
  • Small footprint
  • Low cost, one box solutions
  • Suits all general applications

The Puro™ III T and Geno™ II T utilize well established water purification technologies which you can trust to provide you with the quality of water you require, on request. Call for water from the dispense tap on the front of the machine to obtain the concise water quality you need; when you need it.

The unique reverse osmosis (RO) cartridges have a built-in pre-treatment which only requires changing once per year. It means the water is always in optimum condition as there is no deterioration of quality from an RO which is several years old. The high recovery rate of 37% means the cost of waste water is also kept to a minimum. Rejection rate for particles: >99%. Feedwater pressure: 1.4 - 87 psi.

Application: Both models take potable water to Type 3 or 2 at a rate of 10 or 20 liters per hour which can be dispensed from the 35 liter reservoir. Great for application requiring production of 120 liters or less of RO or DI water.

Certifications: Manufactured to ISO 9001:2015 standard. Certified to IEC 61010-1:2010 and EN 61326-21:2013

Delivery information: All integral pumps and first round of consumables included with unit purchase. Ships within 1 week and installed within 4 to 6 weeks of order.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty