Would you like to log in again?
Epredia™ HistoScreen™ Cassettes, Epredia
Supplier: Epredia
B851001GN B851001WH B851001GY B851000BE B851001BL B851000LI B851001LI B851000GY B851000LA B851001BE B851001OR B851000GN B851000PK B851000TN B851000YW B851000LG B851001TN B851000OR B851001LA B851000WH B851000BL B851001YW B851001PK B851001LG
76337-490CS 1233.52 USD InStock
76337-490 76337-496 76337-486 76337-530 76337-520 76337-498 76337-510 76337-488 76337-492 76337-494 76337-484 76337-526 76337-504 76337-516 76337-528 76337-506 76337-518 76337-522 76337-500 76337-512 76337-502 76337-524 76337-514 76337-508
Epredia™ HistoScreen™ Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
HistoScreen™ cassette mesh screen design accommodates extremely small biopsy specimens.
  • Designed for handling small biopsies and fragile specimens
  • Unique design provides an inner chamber lined with a 'see-through' mesh
  • Eliminates the need for sponges or lens papers
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Variety of 12 colors with matching lids
  • Pore style: Mesh
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty