Would you like to log in again?
Epredia™ PrintMate™ Embedding Cassettes in Tubes, Epredia
Supplier: Epredia
B851100LG B851100PK B851010BE B851100GY B851100GN B851100TN B851100BL B851010TN B851010OR B851100OR B851010LI B851100BE B851100LI B851010WH B851010PK B851100LA B851010GY B851010LA B851010YW B851010LG B851010BL B851100WH B851100YW B851010GN
76337-540CS 525.46 USD InStock
76337-540 76337-672 76337-662 76337-552 76337-652 76337-542 76337-674 76337-532 76337-664 76337-554 76337-670 76337-550 76337-660 76337-658 76337-548 76337-538 76337-654 76337-544 76337-666 76337-534 76337-656 76337-546 76337-668 76337-536
Epredia™ PrintMate™ Embedding Cassettes in Tubes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate embedding cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes packed in plastic tubes (sleeves) for PrintMate™ Printer compatibility
  • With or without matching lids
  • Pore style: Round holes
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty