Would you like to log in again?
Epredia™ PrintMate™ Embedding Cassettes, Epredia
Supplier: Epredia
B851040WH B851070LA B851070LI B851070LG B851070YW B851040BE B851040GY B851040LA B851040TN B851040BL B851040PK B851070GY B851040YW B851070PK B851070GN B851070TN B851070WH B851070BE B851070BL B851040LI B851040OR B851040LG B851040GN B851070OR
76337-590CS 644.35 USD InStock
76337-590 76337-562 76337-584 76337-574 76337-596 76337-564 76337-586 76337-576 76337-598 76337-580 76337-592 76337-570 76337-582 76337-560 76337-572 76337-594 76337-588 76337-566 76337-556 76337-578 76337-600 76337-568 76337-602 76337-558
Epredia™ PrintMate™ Embedding Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate embedding cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Loose bulk packaging configuration of 1000 cassettes
  • With or without matching lids
  • Pore style: Round holes
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty