Would you like to log in again?
Epredia™ PrintMate™ Biopsy Cassettes, Epredia
Supplier: Epredia
B851110WH B851110LG B851110LA B851110BE B851080OR B851080YW B851080LA B851110BL B851080GY B851110YW B851110LI B851080BL B851080TN B851110TN B851080GN B851080WH B851080LG B851080LI B851110OR B851110GY B851080PK B851110PK B851110GN B851080BE
76337-694CS 525.42 USD InStock
76337-694 76337-684 76337-696 76337-686 76337-620 76337-690 76337-680 76337-692 76337-682 76337-614 76337-626 76337-604 76337-616 76337-606 76337-676 76337-610 76337-698 76337-688 76337-622 76337-612 76337-678 76337-624 76337-618 76337-608
Epredia™ PrintMate™ Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate biopsy cassettes are designed for ease of use and print clarity.
  • For small biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty