Would you like to log in again?
Epredia™ PrintMate™ Slotted Cassettes, Epredia
Supplier: Epredia
B851120YW B851120BL B851090WH B851090PK B851090LI B851090LG B851090BE B851090OR B851120TN B851120BE B851090BL B851120LI B851120GY B851090GN B851120LG B851090TN B851120OR B851120GN B851120LA B851120PK B851120WH B851090GY B851090YW B851090LA
76337-650CS 644.35 USD InStock
76337-650 76337-640 76337-630 76337-642 76337-702 76337-636 76337-648 76337-714 76337-638 76337-704 76337-628 76337-716 76337-632 76337-720 76337-644 76337-710 76337-722 76337-700 76337-634 76337-712 76337-646 76337-706 76337-718 76337-708
Epredia™ PrintMate™ Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate slotted cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty