Would you like to log in again?
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes, Epredia
Supplier: Epredia
B851720LG B851954BE B851954TN B851954LA B851954YW B851720WH B851720PK B851720GN B851954BL B851720BE B851720LA B851720TN B851954WH B851954GY B851954GN B851720BL B851954LG B851720OR B851954OR B851720YW B851954PK B851720GY B851720LI B851954LI
76338-006CS 346.12 USD InStock
76338-006 76338-004 76338-008 76337-992 76337-740 76337-994 76337-730 76337-990 76337-988 76337-724 76337-746 76337-736 76337-726 76337-738 76337-742 76337-996 76337-732 76337-744 76337-734 76337-998 76338-010 76337-728 76338-002 76338-000
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
TissueLoc™ Cassette IV slotted cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Large lid tab for easy opening
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty