Would you like to log in again?
Epredia™ Cassette II Slotted Cassettes, Epredia
Supplier: Epredia
B851729TN B851739YW B851739GN B851729BL B851729GN B851729WH B851739GY B851739WH B851729LI B851739PK B851739LI B851729BE B851739LG B851729LG B851729YW B851729OR B851739BL B851729GY B851729LA B851729PK B851739LA B851739TN B851739BE B851739OR
76337-760CS 226.19 USD InStock
76337-760 76337-750 76337-762 76337-752 76337-770 76337-768 76337-922 76337-758 76337-934 76337-748 76337-924 76337-936 76337-764 76337-754 76337-930 76337-766 76337-920 76337-932 76337-756 76337-926 76337-938 76337-916 76337-928 76337-918
Epredia™ Cassette II Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Cassette II slotted cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty