Would you like to log in again?
Epredia™ Thinline Slotted Cassettes, Epredia
Supplier: Epredia
B851736OR B851736YW B851736GN B851736PK B851736BL B851736WH B851736LG B851736LI B851736LA B851736BE B851736TN B851736GY
76337-856CS 586.87 USD InStock
76337-856 76337-846 76337-858 76337-848 76337-852 76337-864 76337-854 76337-866 76337-844 76337-860 76337-850 76337-862
Epredia™ Thinline Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Thinline slotted cassettes come with attached lids to ease of use during handling.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes in plastic tubes (sleeves) for PrintMate™ Printer compatibility
  • Includes matching lids attached
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty