Would you like to log in again?
Epredia™ Thinline Biopsy Cassettes, Epredia
Supplier: Epredia
B851737BE B851737YW B851737BL B851737PK B851737TN B851737LA B851737GY B851737LG B851737OR B851737LI B851737GN B851737WH
76337-878CS 518.47 USD InStock
76337-878 76337-868 76337-874 76337-886 76337-876 76337-888 76337-870 76337-882 76337-872 76337-884 76337-890 76337-880
Epredia™ Thinline Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Thinline Biopsy Cassettes are designed for ease of use and print clarity.
  • For biopsy tissue specimens
  • Variety of 12 colors
  • Comes in tubes (sleeves) for PrintMate™ Printer compatibility
  • Includes matching lids attached
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty