Would you like to log in again?
Epredia™ Cassette II Biopsy Cassettes, Epredia
Supplier: Epredia
B851730LI B851738GN B851738BL B851738OR B851730TN B851730BE B851738LI B851730PK B851738WH B851738YW B851738PK B851730LA B851730YW B851738LG B851730BL B851730WH B851730LG B851738BE B851730OR B851738GY B851738TN B851738LA B851730GY B851730GN
76337-782CS 226.36 USD InStock
76337-782 76337-892 76337-772 76337-794 76337-784 76337-894 76337-774 76337-790 76337-780 76337-792 76337-900 76337-912 76337-902 76337-914 76337-896 76337-786 76337-776 76337-788 76337-898 76337-910 76337-778 76337-908 76337-904 76337-906
Epredia™ Cassette II Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Cassette II biopsy cassettes are designed for ease of use and print clarity.
  • For biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty