Would you like to log in again?
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes, Epredia
Supplier: Epredia
B851750LG B851925LG B851750WH B851925GN B851925BE B851925BL B851925PK B851750GN B851750LI B851750GY B851750BE B851925WH B851925YW B851750OR B851750YW B851750TN B851925OR B851925GY B851925LA B851925TN B851750PK B851750BL B851750LA B851925LI
76337-980CS 282.06 USD InStock
76337-980 76337-970 76337-960 76337-982 76337-950 76337-972 76337-966 76337-944 76337-978 76337-956 76337-968 76337-946 76337-958 76337-940 76337-984 76337-962 76337-974 76337-952 76337-964 76337-986 76337-942 76337-954 76337-976 76337-948
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
TissueLoc™ Cassette IV biopsy cassettes are designed for ease of use and print clarity.
  • For small biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Large lid tab for easy opening
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
Sectioning uses precision engineering for solid connection to microtomes. Cassette surface texture optimizes scanability for archiving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty