Would you like to log in again?
Perforated Basket, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00368229-38
76338-120EA 719.32 USD InStock
76338-120
Perforated Basket, Marlin Steel Wire Products
Baskets Wire Baskets
Rectangular material handling basket with handles.
  • Natural finish
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty