Would you like to log in again?
Hei-VAP Expert Rotary Evaporators, Heidolph®
Supplier: Heidolph
36041016 36041018 36041015 36041005 36041014 36041007 36041009 36041008 36041013 36041011 36041012 36041017 36041000 36041010 36041006 36041001 36041002 36041019 36041004 36041003
76338-336EA 12996.96 USD InStock
76338-336 76338-358 76338-348 76338-326 76338-356 76338-334 76338-346 76338-324 76338-338 76338-328 76338-350 76338-340 76338-354 76338-332 76338-344 76338-322 76338-352 76338-330 76338-342 76338-320
Hei-VAP Expert Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
  • Clear display of all values
  • Central operating panel with 5" touch display
  • Separate knobs for rotation and heating with locking function to prevent accidental readjusting
  • LED ring lights and pictogram to display the activity even from a distance
  • USB and Micro SD interfaces
  • Optional display of vapor temperature
  • Subsequent upgrade to Hei-VAP Expert Control possible, including vacuum and chiller control
It has integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller with timer, limitation of maximum temperature, multilingual menu and text display for error messages. Available in Hand Lift (HL) and Motor Lift (ML) Configurations.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty