Would you like to log in again?
Hei-VAP Ultimate Rotary Evaporators, Heidolph®
Supplier: Heidolph
36041043 36041058 36041041 36041059 36041050 36041055 36041054 36041053 36041040 36041047 36041045 36041046 36041051 36041042 36041049 36041044 36041057 36041048 36041052 36041056
76338-402EA 11826.36 USD InStock
76338-402 76338-424 76338-436 76338-414 76338-400 76338-422 76338-412 76338-434 76338-406 76338-428 76338-418 76338-426 76338-404 76338-438 76338-416 76338-408 76338-420 76338-410 76338-432 76338-430
Hei-VAP Ultimate Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
  • Clear display of all values
  • Central operating panel with 7" touch display
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • LED ring lights and pictogram to display the activity even from a distance
  • USB and Micro SD plus LAN / RS 232 interfaces
  • Optional display of vapor temperature
  • Define and save own evaporation processes (ramps)
It has Separate knobs for rotation and heating with locking function to prevent accidental readjusting, with integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller. Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty