Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oakton® pH 550 Benchtop pH Meter Kits, Cole-Parmer®

Supplier: Cole Parmer

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

35419-30
76338-508EA 613.51 USD InStock
76338-508
Oakton® pH 550 Benchtop pH Meter Kits, Cole-Parmer®
Electrochemical Meters pH and Multi-Parameter Meters
Economical meter for benchtop pH measurement.

  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP 54-rated housing is dust- and spill-proof

The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.

The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.

Certifications: CE compliant.

Accessories information: 550 Series Benchtop Meter Accessories – choose from a wide range of electrodes for your application. Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.

Delivery information: Supplied with Plastic 3-in-1 combination pH electrode, electrode holder, pH standard buffer solutions (4.00/7.00/10.01 pH, 50 ml each), and 9 VAC power adapter.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty