Would you like to log in again?
Oakton® Conductivity and TDS Standards, Cole Parmer
Supplier: Cole Parmer
Oakton®
00653-36 00653-15 00653-18 00653-27 00653-32 00653-20 00653-16 00653-35 00653-23 00653-37 00653-47 00653-89 00653-38 WD-00653-37 00653-50
76338-534EA 19.15 USD InStock
76338-534 76338-556 76338-546 76338-554 76338-544 76338-538 76338-536 76338-558 76338-548 76338-560 76792-334 76338-552 76338-542 76338-550 76338-540
Oakton® Conductivity and TDS Standards, Cole Parmer
Standards Conductivity Standards
Ideal for laboratory and field analysis.
  • Traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25 °C
Oakton conducitivity and TDS standards feature temperature corrections, KCl, NaCL, and 442 conversions printed directly on the bottle for ease of use (excluded on 60-mL bottle, but included in insert).
Certifications: These standards are manufactured under ISO 9000 quality guidelines and traceable to NIST standards.
Caution: If unopened, shelf life is one year. If opened, shelf life if 4 months.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty