Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNext Companion Module for Oxford Nanopore Technologies Ligation Sequencing, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7180S E7180S E7180L
76339-730EA 986 USD InStock
76339-730 CA76339-730 76559-408
NEBNext Companion Module for Oxford Nanopore Technologies Ligation Sequencing, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext DNA repair, end repair and ligation reagents recommended in Ligation library preparation are now available in the same product, at volumes designed for use in several protocols.
  • Component volumes tailored for use with SQK-LSK109
  • Simplified ordering and inventory management
  • Compatible with all devices: MinION, GridION, PromethION, Flongle
  • No more waste - no unnecessary buffers or excess reagents
The appropriate device-specific SQK-LSK109 protocol from Oxford Nanopore Technologies should be followed for use of these reagents.
Delivery information: This module contains the following reagents, at volumes designed for use alongside Oxford Nanopore Technologies SQK-LSK109: NEBNext FFPE DNA Repair Mix (0.048 ml), NEBNext FFPE DNA Repair Buffer (0.084 ml), NEBNext Ultra II End Prep Enzyme Mix (0.072 ml), NEBNext Ultra II End Prep Reaction Buffer (0.084 ml) and Quick T4 DNA Ligase (0.240 ml).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty