Would you like to log in again?
Quick CIP
Supplier: New England Biolabs
M0525S M0525L M0525S M0525L
76339-742EA 478 USD InStock
76339-742 76339-744 CA76339-742 CA76339-744
Quick CIP
Enzymes
Quick CIP is a heat-labile recombinant version of calf intestinal alkaline phosphatase (CIP) purified from a recombinant source. Quick CIP nonspecifically catalyzes the dephosphorylation of 5 and 3 ends of DNA and RNA phosphomonoesters. It also hydrolyses ribo- and deoxyribonucleoside triphosphates (NTPs and dNTPs).
  • Rapid and irreversible heat inactivation eliminates unwanted activity
  • Improved storage stability versus native enzyme
  • Faster reaction setup (no supplemental additives like zinc required) and shorter incubation time
  • Flexible reaction conditions (active in any restriction enzyme buffer, no clean-up required)
Quick CIP is useful in many applications that require the dephosphorylation of DNA or RNA ends. In cloning, dephosphorylation prevents re-ligation of linearized plasmid DNA. The enzyme can quickly dephosphorylate 5 protruding, 5 recessed, and blunt ends in just 10 minutes. Quick CIP may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty