Would you like to log in again?
EnGen Sau Cas9
Supplier: New England Biolabs
M0654S M0654S M0654T M0654T
76339-752EA 158 USD InStock
CA76339-750 76339-752 CA76339-752 76339-750
EnGen Sau Cas9
Enzymes
EnGen Sau Cas9 from Staphylococcus aureus is an RNA-guided DNA endonuclease that catalyzes site-specific cleavage of double-stranded DNA.
  • Longer 5' NNGRRT 3' PAM sequence allows for higher specificity
  • Two nuclear localization signals for improved transport into the nucleus
  • High concentration liquid format option
  • Compatible with the EnGen mutation detection kit
Targeting requires a ~100 nucleotide guide RNA (gRNA) complementary to the target site, as well as a 5' NNGRRT 3' (N = any, R = A or G) protospacer adjacent motif (PAM) on the DNA strand opposite the target sequence. DNA cleavage by EnGen Sau Cas9 occurs ~3 bases 5' of the PAM. EnGen Sau Cas9 has Simian virus 40 (SV40) T antigen nuclear localization signals (NLS) at both the N- and C-termini of the protein. Schematic representation of Sau Cas9 nuclease sequence recognition and DNA cleavage.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty