Would you like to log in again?
Tryptic Soy Agar (TSA) with Lecithin and Tween® 80, Hardy Diagnostics
Supplier: Hardy Diagnostics
U417
76339-810PK 138.59 USD InStock
76339-810
Tryptic Soy Agar (TSA) with Lecithin and Tween® 80, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For use as a general growth medium for the isolation and cultivation of microorganisms. This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms.
  • A crystal clear glass construction provides complete visibility
  • Features a streamlined design with no handle, making it easier to store and clean
  • Easy-to-remove, perforated, shrink wrap ensures product integrity
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty