Would you like to log in again?
AlphaTec® 53-001 Series Gloves, Ansell
Supplier: Ansell
53001080-NA 53001100-NA 53001090-NA 53001070-NA 53001110-NA
76339-862CS 575.86 USD InStock
76339-862 76339-874 76339-854 76339-858 76339-860
AlphaTec® 53-001 Series Gloves, Ansell
Gloves
One glove with the benefits of two, offering multi-barrier chemical protection.
  • Innovative multi-layer polymer design: nitrile/neoprene/nitrile
  • Provides protection against a wide range of chemicals, from acids and bases to hydrocarbons and organic solvents
  • Tested according to EN ISO 374:2016; permeation time >30 minutes against 13 chemicals out of 18 listed
  • Features the latest MICROCHEM™ chemical barrier technology, providing superior protection for use in hazardous environments
  • ANSELL GRIP™ technology provides enhanced dexterity, grip, and comfort when handling wet or oily items
  • Inner soft nylon liner for comfort and increased mechanical protection
The Ansell AlphaTec® 53-001 is a 'two-in-one' nitrile and neoprene multilayer glove that provides broader chemical protection, helps reduce the number of SKUs used in a workplace, and ensures peace of mind for safety managers and workers in hazardous chemical applications.
Certifications: EN388:2016 3121A; EN407 X1XXXX; EN420:2003 + A1:2009; EN ISO 374-1:2016 Type A ACFGJKLMNOPST.
Packaging: Gloves are packaged 6 pairs per bag, 12 bags per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty