Would you like to log in again?
Limited Flammability Economy Safety Vest, VCL2MLFR Series, ANSI/ISEA Class 2, MCR Safety
Supplier: MCR Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VCL2MLFRX3 VCL2MLFRL VCL2MLFRX4 VCL2MLFRX2 VCL2MLFRM VCL2MLFRXL
76340-004EA 25.72 USD InStock
76340-004 76340-246 76340-166 76340-132 76340-050 76340-006
Limited Flammability Economy Safety Vest, VCL2MLFR Series, ANSI/ISEA Class 2, MCR Safety
Workwear
These fluorescent lime safety vests are ideally suited for applications requiring high visibility apparel.
  • Hook and loop front closure
  • 2" silver reflective stripes
  • Two outer pockets, one inner pocket
  • NOT designed for protection against ARC Flash or Flash Fire
Mesh construction provides increased breathability in warm environments.
Certifications: ANSI/ISEA 107-2015 Class 2, Type R compliant.
Caution: Limited flammability protection. This garment is not flame resistant, as defined by ANSI/ISEA 107-2015 Section 10.5.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty