Would you like to log in again?
Wizard Series PVC/Nylon/PVC Rain Jackets with Detachable Hood, MCR Safety
Supplier: MCR Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
300JXL 300JX4 300JX3 300JL 300JX2 300JX5 300JM
76341-150EA 42.57 USD InStock
76341-150 76341-064 76341-008 76341-114 76341-038 76340-874 76340-952
Wizard Series PVC/Nylon/PVC Rain Jackets with Detachable Hood, MCR Safety
Workwear
This wizard series coat features a 0.28 mm double coating of PVC, with tough nylon scrim material layered in between.
  • Equipped with detachable drawstring hood
  • Take up snaps on the wrists
  • Storm flap under snap front closure
  • Rip stop scrim material provides excellent resistance to snags and tears
Nylon scrim has excellent rip and puncture resistance. This yellow two-piece jacket is a waist length option with a detachable drawstring hood and storm flap under a snap up front closure. Limited flammability, this garment is not intended for use as protection against ARC flash or flash fire.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty