Would you like to log in again?
Steel Toe Boots, 16" PVC, with Cleated Sole and Polyester Liner, MCR Safety
Supplier: MCR Safety
VBS12010 VBS1208 VBS1209 VBS12012 VBS12011 VBS12014 VBS1207 VBS12013
76342-394PR 47.71 USD InStock
76342-394 76342-528 76342-486 76342-574 76342-544 76342-476 76342-524 76342-558
Steel Toe Boots, 16" PVC, with Cleated Sole and Polyester Liner, MCR Safety
Shoes and Boots
Over-the-sock style black PVC boots feature a steel toe and polyester lining for comfort.
  • Steel toe
  • Over-the-sock design
  • Polyester lining for increased comfort
  • Cleated sole and kick-off lug
  • 16" height
Boots feature a deeper cleated sole for improved grip, polyester lining, and a kick-off lug to aid in removal.
Certifications: ASTM F2413 Impact and Compression compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty