Would you like to log in again?
SAFETY GLASSES DM3 2.0 CLEAR MAX6 PK12
Supplier: MCR SAFETY
Glasses Safety Dm3 2.0 Clear Max6 PK12PR^ Dominator* Safety Glasses, DM3, Magnifier 2.0, Frame Material: Polycarbonate, Lens Options:Clear, Lens Coating:MAX6 Anti-Fog, with Scratch Resistant, ANSI Z87+, VLT: 90%, Frame Color:Tortoiseshell, Temple Color:To
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAFETY GLASSES DM3 2.0 CLEAR MAX6 PK12 76342-964 DM13H20PF Pack of 12 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty