Would you like to log in again?
VWR® Clinical 4-Way Tube Racks
Supplier: VWR
VWR®
120687V 120684V 120686V 120683V 120685V
76343-406CS 495.62 USD InStock
76343-406 76343-408 76343-410 76343-412 76343-404
VWR® Clinical 4-Way Tube Racks
Racks Tube Racks
These autoclavable tube racks have a six-way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezer
  • Accommodates all brands of 13- and 17-mm tubes
They can be used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed.
Packaging: Pack of five.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty