Synonyms:
Acetylcysteine
, (R)-2-Acetamido-3-mercaptopropanoic acid
, (R)-(+)-N-Acetylcysteine
, Ac-Cys-OH
A10040-100.0
A10040-25.0
A10040-500.0
A10040-50.0
76344-478EA
89.67
USD
InStock
76344-478
76344-484
76344-482
76344-480
N-Acetyl-L-cysteine ≥98.5%
N-Acetyl-L-cysteine
Increases cellular pools of free radical scavenger
- Prevents apoptotic death of neuronal cells.
- Induces apoptosis in human and rat smooth muscle cells.
- Pyrogen free.