Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bile Salts #3, RPI

Supplier: RPI
B11750-25.0 B11750-500.0 B11750-100.0
76345-090EA 71.6 USD InStock
76345-090 76345-088 76345-092
Bile Salts #3, RPI
Media for Microbiology Additives and Supplements for Microbiology
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate.

It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.

Sodium Cholate: 45.0 - 55.0 %
Sodium Deoxycholate: 45.0 - 55.0 %

Appearance:White Powder
Loss on Drying: ≤ 5.0 %
Solubility (2% in water, autoclaved): Clear

Caution: Research or further manufacturing use only, not for food or drug use.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty