Synonyms:
6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
, α-Carboxybenzylpenicillin disodium salt
C46000-5.0
C46000-0.25
C46000-25.0
C46000-1.0
C46000-100.0
C46000-500.0
76345-522EA
783.45
USD
InStock
76345-522
76345-524
76345-520
76345-518
76345-526
76345-516
Carbenicillin disodium salt ≥92.0% (by HPLC) USP
Carbenicillin disodium salt
USP Grade. Prevents bacterial cell wall synthesis. Inhibits crosslinking of peptidoglycan.
- Highly active against gram negative bacteria
- Commonly used for the elimination of Agrobacterium species after inoculation
- Sensitive to B-Lactamase
- Non-toxic to plant cells
- Plant cell culture tested
- Hygroscopic, soluble in water