Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Luria Broth 1 Liter Buffered Capsules [Miller's LB Broth], RPI

Supplier: RPI
L24045-5000.0 L24045-1000.0 L24045-500.0
76346-566EA 210.25 USD InStock
76346-566 76346-568 76346-570
Luria Broth 1 Liter Buffered Capsules [Miller's LB Broth], RPI
Media for Microbiology Dehydrated Media for Microbiology
RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

  • For maintenance and propagation of E. Coli
  • High salt formula
  • Molecular biology grade

Simply dissolve 1 capsule per 1 Liter of distilled water. Water must be 37 °C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.2 at 20°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

Microorganism (E. coli ATCC 23724): Good to Excellent
Microorganism (E. coli ATCC 33694): Good to Excellent
Microorganism (E. coli ATCC 33849): Fair to Excellent
Microorganism (E. coli ATCC 39403): Good to Excellent
Microorganism (E. coli ATCC 47014): Good to Excellent
Microorganism (E. coli ATCC 53868): Good to Excellent

Caution: Research or further manufacturing use only, not for food or drug use.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty