Would you like to log in again?
SLEEVES STRING CUTPRO 15IN KEVLAR PK10
Supplier: MCR SAFETY
Sleeves String Cutpro 15In Kevlar PK10^ CutPro* Sleeves, Material Group: Strings, Material: Kevlar, Overall Length: 15 in, ANSI Cut: A3, ANSI Abrasion: 2, ANSI Contact Heat: 2 - Less than 392 deg F, Applications: Demolition, Furniture industry, Landscaping, Mechanics
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SLEEVES STRING CUTPRO 15IN KEVLAR PK10 76349-046 9375 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty