Would you like to log in again?
GLOVES 13 GAUGE POLYURETHANE XL 1 PAIR
Supplier: MCR SAFETY
Gloves 13 Gauge Polyurethane Xl 1 PAIR^ Safety Cut Pro* Gloves, 13 Gauge, Material: HyperMax* Shell, HPPE, Series: Memphis, Polyurethane Coated Palm and Fingertips, Outstanding Cut and Abrasion Resistance, Gray PU palm and fingers, Comfortable and easy to wear, Good dexterity, Size: XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES 13 GAUGE POLYURETHANE XL 1 PAIR 76349-364 92743PUXL Pair Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty