Would you like to log in again?
GLOVES NINJA FORCE 13 G PU PALM XL 1PR
Supplier: MCR SAFETY
Gloves Ninja Force 13 G Pu Palm Xl 1PR^ Ninja* Force Gloves, Cut Protection, Material Group: Strings, Material: Dyneema, Gauge: 13 Gauge, PU Palm and Fingertips Coated, Point of Purchase Hang Tag, Applications: Automotive Assembly, Farming, Landscaping, Agriculture, RoofingsizeXL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NINJA FORCE 13 G PU PALM XL 1PR 76349-522 N9677XL Pair Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty