Would you like to log in again?
GLOVES MULTI-TASK FORCEFLEX W/TPR M 1PR
Supplier: MCR SAFETY
Gloves Multi-Task Forceflex W/Tpr M 1PR^ Forceflex* Multi-Task Gloves, Reinforced Clarino* Synthetic Leather Palm, Mesh Back with TPR metacarpal protection, Injection molded TPR padding on the back of hand and fingers, yellow mesh fabric backing, Adjustable hook and loop, size: M
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES MULTI-TASK FORCEFLEX W/TPR M 1PR 76350-174 Y200M Pair Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty