Would you like to log in again?
GLOVES POLYBAG YL LATEX M DZ PR
Supplier: MCR SAFETY
Gloves Polybag Yellow Latex M DZ 1PR^ Gloves, 18 mil Flock Lined yellow Latex, 12in length with straight cuff, Polybag per pair, Component materials comply with all relevant federal regulations for food contact, size: M
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES POLYBAG YL LATEX M DZ PR 76351-124 5280P Dozen(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty