Would you like to log in again?
GLOVES RED RAM GRAIN GOAT S DZ 1PR
Supplier: MCR SAFETY
Gloves Red Ram Grain Goat S DZ12 1PR^ Gloves, Red Ram*, Grain Goat, Straight Thumb, 5in Split Cow Cuff, DuPont* Kevlar* Sewn, Greater softness for sense of touch, Exceptional durability,Grain kid goatskin, MIG/TIG, sewn with DuPont* Kevlar*, size: S
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES RED RAM GRAIN GOAT S DZ 1PR 76352-484 4840S Dozen(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty