Would you like to log in again?
GLOVES MEMPHIS 14 IN GR XXL DZ 1PR
Supplier: MCR SAFETY
Gloves Memphis 14 In Gr Xxl DZ12 1PR^ Memphis NeoMax* Gloves, Green Double Dipped Neoprene, 100% Cotton Brushed Interlock Liner, Treated with Actifresh*, 14 Inch Length, Sandy Finish, Component materials comply with all federal regulations for food contact, size: XXL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES MEMPHIS 14 IN GR XXL DZ 1PR 76353-248 6914XXL Dozen(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty