Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dustless Precision Pellets®, Sucrose, Bio-Serv

Supplier: Bio-Serv
Dustless Precision Pellets®
F0023 F0025 F07595 F0024 F07595 F0024 F0025 F0026 F0023 F0026
76285-258BX 142.21 USD InStock
76285-258 76285-260 76285-262
Dustless Precision Pellets®, Sucrose, Bio-Serv
Animal Supplies Animal Feed
Bio-Serv is now offering an alternative sucrose pellet formulation, which is available unflavored, or in chocolate or banana flavors.

  • Shelf Life: 12 months, cool dry storage

Ingredients: F0023 - Sucrose, Invert Sugar, Magnesium Stearate, Calcium Silicate; F0024 - Sucrose, Invert Sugar, Banana Flavor, Magnesium Stearate, Calcium Silicate, Food Dye; F07256, F0025 - Sucrose, Invert Sugar, Chocolate Flavor, Magnesium Stearate, Calcium Silicate.

Caloric value: 3.84 kcal/gm
Nutritional ash content: 1.5%
Nutritional carbohydrate content: 94.8%
Nutritional moisture content:<5%

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty