Would you like to log in again?
Rat MD's™ Meloxicam Bacon Flavor
Supplier: Bio-Serv
MD275-050 MD275-050-25 MD275-050 MD275-050-25
77506-114PK 40.09 USD InStock
CA77506-114 77506-114 76353-778 CA76353-778
Rat MD's™ Meloxicam Bacon Flavor
Animal Supplies Animal Feed
Meloxicam is an effective anti-inflammatory drug used in a variety of species including rodents.
Meloxicam MD's are highly palatable and encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and injurious post-operative injections.
Meloxicam MD's can be customized to include antibiotics such as Baytril to provide both medications easily and affordably.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty