Would you like to log in again?
Primate Medications, Bio-Serv
Supplier: Bio-Serv
MD950-4 MD925-500 MD800-100 MD975-40 MD275-080 MD500-50 F05235 MD050-50 F05237 MD900-222 MD200-120 MD750-250 MD200-120 MD925-500 MD150-15 MD500-50 MD100-80 MD150-15 MD700-100 MD900-222 MD300-20 MD800-100 MD600-100 MD600-100 MD750-100 MD050-50 MD750-250 MD975-40 F05235 MD350-20 MD750-100 MD300-20 MD275-080 MD100-80 MD350-20 MD950-4 MD700-100 F05237
89067-490PK 178.97 USD InStock
89067-490 89067-508 89408-214 CA89067-488 89067-512 CA89067-500 89408-212 CA89067-502 89408-218 CA89067-504 89408-216 89067-492 89067-496 CA89067-484 89067-498 CA89067-486 CA89067-506 CA89067-508 89408-210 CA89067-498 89067-500 CA89408-212 89067-502 CA89067-512 CA89408-210 89067-504 CA89408-216 89067-506 CA89408-214 CA89067-490 CA89067-492 89067-484 CA89408-218 89067-486 CA89067-496 89067-488
Primate Medications, Bio-Serv
Animal Supplies Animal Feed
Chewable, flavored, medicated 5 g tablet replaces more stressful dosing methods
Base Meal Ingredients: Sucrose, Dextrose, Mineral Oil, Artificial and Natural Flavors, Calcium Silicate, Magnesium Stearate.
Note: A signed MD Form is required for all Medications except Aspirin and L-Tryptophan. Please Fax the completed form to 908-284-4753. The form will remain on file for one year. Refills are permitted as long as the prescribing veterinarian listed on the request form is still affiliated with the requesting institution.
Tablet consistency ensures accurate dosing; scored for flexible dose administration. Cold tableting process eliminates drug degradation. No heat or water used in processing. Highly palatable sugar-based formula with excellent drug-masking capabilities. Reduces time, labor and stress compared to traditional dosing methods. Medications can be customized to meet specific requirements. Shelf Life: 6-24 months as dictated by the drug expiration date.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty