Would you like to log in again?
SD Pro™ Primate Probiotic Tablets, Bio-Serv
Supplier: Bio-Serv
SD Pro™
PB100-BLK-B PB100-30-B PB100-BLK PB100-BLK PB100-BLK-B PB100-30 PB100-30
76285-296BX 1366.51 USD InStock
76285-296 76285-294 CA76353-786 CA10798-916 CA76285-296 76353-786 10798-916
SD Pro™ Primate Probiotic Tablets, Bio-Serv
Animal Supplies Animal Feed
SD Pro™ Primate Probiotic Tablets are a non-human primate origin bacteria probiotic featuring Lactobacillus reuteri.
Ingredients: Sucrose, Dextrose, Strawberry Flavor, Yeast Culture, Hydrolyzed Yeast, Calcium Silicate, Magnesium Stearate, Dried Lactobacillus Reuteri, Fermentation Product, Soybean Oil, Sodium Aluminosilicate.
Feeding Directions: <1 kg primate: ¼ tablet daily; 1-2 kg primate: ½ tablet daily; 3-10 kg primate: 1 tablet daily; >10 kg primate: 2-4 tablets daily. Note: If non-human primate is on antibiotic therapy, administer PrimiOtic 4–6 hours after antibiotic dosage.
L. reuteri is a natural inhabitant of the GI tract and contains antimicrobial properties, which prevents the colonization of pathogenic bacteria. Each tablet contains 1.25 billion CFU of live Lactobacillus reuteri in a highly palatable, easy-to-dose tablet. Restores and maintains gastrointestinal health. Shelf Life: 6 months from date of manufacture. Refrigerated. Primiotic contains live culture, which is sensitive to heat and moisture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty